Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172758.4 | NP_766346.1 | 12 | 2266 | ||
GenBank mRNA | AK052548.1 | - | 12 | 2267 | ||
AK171137.1 | - | 12 | 2274 | |||
AK183874.1 | - | 1 | 221 | |||
BC031853.1 | - | 10 | 2139 | 4009949 |