siRNA Details
Gene Symbol: Ccdc108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039495.1 | NP_001034584.1 | 9 | 1501 | ||
GenBank mRNA | AK038623.1 | - | 7 | 1207 | ||
AK046272.1 | - | 8 | 1312 | |||
AK133278.1 | - | 9 | 1502 | |||
BC150939.1 | - | 9 | 1447 | 9087850 |
