Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253831.1 | NP_001240760.1 | 2 | 296 | ||
NM_001253834.1 | NP_001240763.1 | 3 | 387 | |||
NM_001253836.1 | NP_001240765.1 | 2 | 319 | |||
NM_175025.4 | NP_778190.3 | 3 | 502 | |||
GenBank mRNA | AJ551270.1 | - | 1 | 84 | ||
AK007226.1 | - | 4 | 288 | |||
AK028393.1 | - | 2 | 265 | |||
AK032255.1 | - | 4 | 458 | |||
AK052958.1 | - | 4 | 487 | |||
AK133703.1 | - | 2 | 297 | |||
AK136062.1 | - | 387 | ||||
AK159918.1 | - | 3 | 453 | |||
AK163049.1 | - | 3 | 348 | |||
AK173154.1 | - | 1 | 75 | |||
AK209035.1 | - | 1 | 20 | |||
BC043091.1 | - | 3 | 468 | 6405251 | ||
BC043706.1 | - | 3 | 449 |