Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199632.1 | NP_001186561.1 | 5 | 544 | ||
NM_177076.4 | NP_796050.2 | 5 | 544 | |||
GenBank mRNA | AK029566.1 | - | 5 | 546 | ||
AK030172.1 | - | 5 | 527 | |||
BB614373.1 | - | 5 | 545 | |||
BC145291.1 | - | 5 | 470 | 9053845 | ||
BC145292.1 | - | 5 | 471 | 9053846 |