Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Duxbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183389.1 | NP_899245.1 | 3 | 741 | ||
GenBank mRNA | AK134879.1 | - | 910 | |||
AK141686.1 | - | 3 | 417 | |||
BC055782.1 | - | 3 | 741 | 6403934 | ||
EF472598.1 | - | 3 | 478 |
Gene Symbol: Duxbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177538.1 | NP_001171009.1 | 3 | 731 | ||
GenBank mRNA | EU257807.1 | - | 3 | 315 |
Gene Symbol: Duxbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177539.1 | NP_001171010.1 | 731 |