Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008466.5 | NP_032492.1 | 1, 2 | 235 | ||
GenBank mRNA | AF020772.1 | - | 1, 2 | 79 | ||
AK011559.1 | - | 1, 2 | 106 | |||
AK049373.1 | - | 1, 2 | 106 | |||
AK161664.1 | - | 1, 2 | 163 | |||
AV581906.1 | - | 1, 2 | 235 | |||
BC026885.1 | - | 1, 2 | 129 | 3979788 | ||
CJ063714.1 | - | 1, 2 | 320 |