Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRCAM SIRNA Assays ›
Gene Symbol:
NRCAM
Gene Name:
neuronal cell adhesion molecule
Gene Aliases:
-
Chromosome Location:
Chr.7: 108147626 - 108456442 on Build GRCh38
UniGene ID:
Hs.21422
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
15520
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRCAM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037132.2
NP_001032209.1
4
448
NM_001193582.1
NP_001180511.1
7
828
NM_001193583.1
NP_001180512.1
7
828
NM_001193584.1
NP_001180513.1
7
828
NM_005010.4
NP_005001.3
6
810
XM_005250373.2
XP_005250430.1
7
828
XM_005250383.2
XP_005250440.1
7
828
XM_005250385.2
XP_005250442.1
7
828
XM_006716003.1
XP_006716066.1
7
828
XM_006716007.1
XP_006716070.1
7
828
XM_006716012.1
XP_006716075.1
7
828
XM_006716014.1
XP_006716077.1
7
828
XM_011516253.1
XP_011514555.1
7
828
XM_011516255.1
XP_011514557.1
6
583
XM_011516256.1
XP_011514558.1
7
828
XM_011516257.1
XP_011514559.1
6
810
XM_011516258.1
XP_011514560.1
5
565
XM_011516259.1
XP_011514561.1
7
828
XM_011516261.1
XP_011514563.1
6
810
XM_011516262.1
XP_011514564.1
7
828
XM_011516265.2
XP_011514567.1
7
828
XM_011516266.2
XP_011514568.1
7
828
XM_011516267.1
XP_011514569.1
7
828
XM_011516268.2
XP_011514570.1
5
643
XM_011516269.1
XP_011514571.1
7
828
XM_011516270.1
XP_011514572.1
7
828
XM_011516271.1
XP_011514573.1
6
810
XM_017012236.1
XP_016867725.1
7
828
XM_017012237.1
XP_016867726.1
7
828
XM_017012238.1
XP_016867727.1
7
828
XM_017012239.1
XP_016867728.1
6
810
XM_017012240.1
XP_016867729.1
7
828
XM_017012241.1
XP_016867730.1
6
552
XM_017012242.1
XP_016867731.1
6
552
XM_017012243.1
XP_016867732.1
7
828
XM_017012244.1
XP_016867733.1
7
828
XM_017012245.1
XP_016867734.1
7
828
XM_017012246.1
XP_016867735.1
7
828
XM_017012247.1
XP_016867736.1
7
828
XM_017012248.1
XP_016867737.1
7
828
XM_017012249.1
XP_016867738.1
7
828
XM_017012250.1
XP_016867739.1
6
552
XM_017012251.1
XP_016867740.1
7
828
XM_017012252.1
XP_016867741.1
7
828
XM_017012253.1
XP_016867742.1
4
497
XM_017012254.1
XP_016867743.1
7
828
XM_017012255.1
XP_016867744.1
5
595
XM_017012256.1
XP_016867745.1
4
497
XM_017012257.1
XP_016867746.1
6
583
XM_017012258.1
XP_016867747.1
6
552
XM_017012259.1
XP_016867748.1
4
497
GenBank mRNA
AB002341.1
-
7
756
AJ001057.1
-
4
327
AK299870.1
-
7
816
BC098401.1
-
6
798
30528526
BC114570.1
-
7
752
40083055
BC115736.1
-
7
752
40083062
BX538010.1
-
6
755
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
neuron migration
axonogenesis
axonal fasciculation
synapse assembly
central nervous system development
protein localization
single organismal cell-cell adhesion
neuronal action potential propagation
regulation of axon extension
retinal ganglion cell axon guidance
heterotypic cell-cell adhesion
clustering of voltage-gated sodium channels
positive regulation of neuron differentiation
protein binding
ankyrin binding
protein binding involved in heterotypic cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-zp8kv:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0