Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195046.1 | NP_001181975.1 | 5, 6 | 559 | ||
NM_001195047.1 | NP_001181976.1 | 4, 5 | 514 | |||
NM_001195048.1 | NP_001181977.1 | 5, 6 | 669 | |||
NM_001195049.1 | NP_001181978.1 | 7, 8 | 933 | |||
NM_008778.3 | NP_032804.2 | 4, 5 | 542 | |||
GenBank mRNA | AF082297.1 | - | 3, 4 | 456 | ||
AJ496262.1 | - | 3, 4 | 424 | |||
AJ496263.1 | - | 4, 5 | 469 | |||
AK031853.1 | - | 5, 6 | 550 | |||
AM943853.1 | - | 4, 5 | 487 | |||
AM943854.1 | - | 5, 6 | 532 | |||
BC053403.1 | - | 4, 5 | 549 | 30060082 | ||
U39738.1 | - | 7, 8 | 940 |