Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC053031.1 | - | 5 | 1057 | 6401146 | |
BC062205.1 | - | 4 | 832 | 4950774 | ||
BC096741.1 | - | 6 | 1035 | |||
D13867.1 | - | 5 | 679 |