Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008002.4 | NP_032028.1 | 2 | 1079 | ||
GenBank mRNA | AK034742.1 | - | 2 | 645 | ||
AK045267.1 | - | 2 | 1080 | |||
AK085244.1 | - | 2 | 401 | |||
AK141633.1 | - | 2 | 401 | |||
BC048229.1 | - | 2 | 744 | 6313081 | ||
D89080.1 | - | 2 | 394 | |||
U94517.1 | - | 2 | 394 |