Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289533.1 | NP_001276462.1 | 6 | 1362 | ||
NM_010212.4 | NP_034342.1 | 6 | 1511 | |||
GenBank mRNA | AF153340.1 | - | 5 | 1278 | ||
AK133708.1 | - | 6 | 1481 | |||
AK147024.1 | - | 6 | 1512 | |||
U77040.2 | - | 6 | 1401 |