Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042707.2 | NP_001036172.1 | 3 | 371 | ||
NM_001042708.2 | NP_001036173.1 | 4 | 410 | |||
NM_001042709.2 | NP_001036174.1 | 3 | 371 | |||
NM_001277321.1 | NP_001264250.1 | 4 | 410 | |||
NM_001277322.1 | NP_001264251.1 | 5 | 338 | |||
NM_010561.3 | NP_034691.2 | 4 | 410 | |||
GenBank mRNA | AB221609.1 | - | 2 | 86 | ||
AF098967.1 | - | 5 | 287 | |||
AF497751.2 | - | 4 | 278 | |||
AF497752.2 | - | 3 | 239 | |||
AK048096.1 | - | 3 | 301 | |||
AK077560.1 | - | 4 | 336 | |||
AK128936.1 | - | 4 | 343 | |||
AK150276.1 | - | 4 | 359 | |||
BB851524.1 | - | 3 | 374 | |||
BC047272.1 | - | 4 | 330 | 6440312 | ||
BC060599.1 | - | 2 | 74 | |||
DQ104405.1 | - | 4 | 278 | |||
DQ104406.1 | - | 3 | 239 |