Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inppl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122739.1 | NP_001116211.1 | 13 | 1629 | ||
NM_010567.2 | NP_034697.2 | 13 | 1863 | |||
GenBank mRNA | AF162781.1 | - | 13 | 1713 | ||
AK133359.1 | - | 13 | 1590 | |||
AK149873.1 | - | 13 | 1647 | |||
BC063080.1 | - | 13 | 1559 | 6416946 |