Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077264.1 | NP_001070732.1 | 12 | 1646 | ||
NM_007458.2 | NP_031484.1 | 12 | 1646 | |||
GenBank mRNA | AK135424.1 | - | 12 | 1575 | ||
AK155515.1 | - | 12 | 1583 | |||
AK198323.1 | - | 2 | 81 | |||
AK201691.1 | - | 81 | ||||
BC031433.1 | - | 12 | 1596 | 4481643 | ||
BC114386.1 | - | 1 | 386 |