Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF012709.1 | - | 5 | 972 | ||
AK011399.1 | - | 5 | 917 | |||
AK053763.1 | - | 5 | 941 | |||
BC011038.1 | - | 5 | 952 | 4018429 | ||
BQ748418.1 | - | 1 | 76 |