Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007685.2 | NP_031711.1 | 6 | 914 | ||
GenBank mRNA | BC100705.1 | - | 6 | 872 | 40044593 | |
BC100706.1 | - | 6 | 872 | 40044594 | ||
BC100708.1 | - | 6 | 872 | 40044595 | ||
BC100711.1 | - | 6 | 872 | 40044596 | ||
U57720.1 | - | 6 | 914 |