Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007742.3 | NP_031768.2 | 19, 20 | 1364 | ||
GenBank mRNA | AK132180.1 | - | 19, 20 | 1383 | ||
AK158011.1 | - | 19, 20 | 1381 | |||
AK159283.1 | - | 19, 20 | 1382 | |||
AK159285.1 | - | 19, 20 | 1384 | |||
BC042642.1 | - | 12, 13 | 934 | |||
BC050014.1 | - | 19, 20 | 1364 | 4225038 | ||
BC059281.1 | - | 19, 20 | 1374 | 3992363 |