Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf3r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252651.1 | NP_001239580.1 | 5 | 651 | ||
NM_007782.3 | NP_031808.2 | 5 | 651 | |||
GenBank mRNA | AK089305.1 | - | 5 | 596 | ||
AK144318.1 | - | 4 | 552 | |||
BC115865.1 | - | 4 | 528 | 40081337 | ||
BC116635.1 | - | 4 | 528 | 40081333 | ||
M58288.1 | - | 5 | 574 |