Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp17a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007809.3 | NP_031835.3 | 7 | 1365 | ||
GenBank mRNA | AK132830.1 | - | 7 | 1259 | ||
AK134376.1 | - | 7 | 1259 | |||
BC064793.1 | - | 7 | 1339 | 30436278 | ||
M64863.1 | - | 7 | 1238 |