Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013811.3 | NP_038839.2 | 728 | |||
GenBank mRNA | AF356520.1 | - | 728 | |||
AF356521.1 | - | 728 | ||||
AF356522.1 | - | 738 | ||||
AF356523.1 | - | 738 | ||||
AF363577.1 | - | 738 | ||||
AK161258.1 | - | 730 |