Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276684.1 | NP_001263613.1 | 2, 3 | 2137 | ||
NM_018790.3 | NP_061260.1 | 2, 3 | 2140 | |||
GenBank mRNA | AF162777.1 | - | 2, 3 | 2131 | ||
AK157822.1 | - | 2, 3 | 2139 | |||
BC023127.1 | - | 2, 3 | 2126 | 3498057 |