Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021272.3 | NP_067247.1 | 4 | 557 | ||
GenBank mRNA | AK002955.1 | - | 4 | 558 | ||
AK021271.1 | - | 4 | 562 | |||
BC055280.1 | - | 4 | 540 | 5710017 | ||
BC057090.1 | - | 4 | 543 | 5700428 | ||
S69799.1 | - | 4 | 539 |