Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272057.1 | NP_001258986.1 | 14 | 1769 | ||
NM_001272058.1 | NP_001258987.1 | 14 | 1769 | |||
NM_001272059.1 | NP_001258988.1 | 14 | 1769 | |||
NM_007401.3 | NP_031427.2 | 14 | 1769 | |||
GenBank mRNA | AK016574.1 | - | 14 | 1425 | ||
AK029519.1 | - | 14 | 1459 | |||
AK161262.1 | - | 14 | 1769 | |||
AK161415.1 | - | 11 | 1147 | |||
AK161417.1 | - | 14 | 1439 | |||
BC094237.1 | - | 14 | 1466 | 30604517 | ||
KF144380.1 | - | 2 | 170 | |||
KF144382.1 | - | 2 | 170 | |||
U22059.1 | - | 14 | 1433 |