Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ostf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017375.3 | NP_059071.1 | 10 | 1047 | ||
GenBank mRNA | AK002899.1 | - | 10 | 1026 | ||
AK005525.1 | - | 10 | 1016 | |||
AK010074.1 | - | 10 | 1043 | |||
AK149175.1 | - | 1025 | ||||
AK150145.1 | - | 10 | 1017 | |||
AK155028.1 | - | 10 | 1017 | |||
AK205236.1 | - | 1 | 104 | |||
AK207431.1 | - | 1 | 185 | |||
AK209921.1 | - | 1 | 185 | |||
AK210606.1 | - | 1 | 185 | |||
AK212936.1 | - | 1 | 185 | |||
BC060986.1 | - | 10 | 926 | 30300544 | ||
U58888.1 | - | 10 | 995 |