Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019821.2 | NP_062795.2 | 5 | 889 | ||
GenBank mRNA | AK003218.1 | - | 1 | 356 | ||
AK157313.1 | - | 5 | 876 | |||
AK185281.1 | - | 1 | 180 | |||
BC016584.1 | - | 5 | 889 | 3157800 | ||
BC053729.1 | - | 5 | 832 | 30065192 |