Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135729.1 | NP_001129201.1 | 5 | 466 | ||
NM_001135732.1 | NP_001129204.1 | 5 | 539 | |||
NM_005488.2 | NP_005479.1 | 5 | 539 | |||
NR_024194.1 | 5 | 568 | ||||
NR_024195.1 | 4 | 489 | ||||
XM_011529818.2 | XP_011528120.1 | 6 | 553 | |||
XM_011529820.2 | XP_011528122.1 | 4 | 404 | |||
XM_017028529.1 | XP_016884018.1 | 6 | 553 | |||
XM_017028530.1 | XP_016884019.1 | 4 | 404 | |||
GenBank mRNA | AJ006973.1 | - | 5 | 475 | ||
AK026576.1 | - | 5 | 492 | |||
AK097926.1 | - | 6 | 555 | |||
AK293694.1 | - | 5 | 466 | |||
AK295918.1 | - | 4 | 399 | |||
AK296670.1 | - | 5 | 476 | |||
AK314522.1 | - | 5 | 448 | |||
AY358980.1 | - | 5 | 474 | |||
BC040006.1 | - | 5 | 432 | |||
BC046151.1 | - | 5 | 417 | 6470390 | ||
CR456595.1 | - | 5 | 435 | |||
DC297234.1 | - | 5 | 539 |