Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Riok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024242.3 | NP_077204.2 | 2, 3 | 753 | ||
GenBank mRNA | AK017312.1 | - | 2, 3 | 448 | ||
AK152707.1 | - | 2, 3 | 502 | |||
AK161315.1 | - | 3, 4 | 996 | |||
BC006884.1 | - | 2, 3 | 460 |