Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178091.1 | NP_001171562.1 | 2, 3 | 613 | ||
NM_001178092.1 | NP_001171563.1 | 1, 2 | 541 | |||
NM_001178093.1 | NP_001171564.1 | 2, 3 | 209 | |||
NM_001178094.1 | NP_001171565.1 | 2, 3 | 506 | |||
NM_005504.6 | NP_005495.2 | 2, 3 | 613 | |||
XM_017019768.1 | XP_016875257.1 | 2, 3 | 332 | |||
GenBank mRNA | AK056255.1 | - | 2, 3 | 613 | ||
AK128527.1 | - | 1, 2 | 168 | |||
AK294879.1 | - | 2, 3 | 192 | |||
AK299088.1 | - | 2, 3 | 197 | |||
BX647122.1 | - | 2, 3 | 215 | |||
CR749308.1 | - | 2, 3 | 415 | |||
DC385021.1 | - | 2, 3 | 210 | |||
U21551.1 | - | 1, 2 | 80 |