Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025982.4 | NP_080258.1 | 4 | 494 | ||
GenBank mRNA | AK012457.1 | - | 4 | 495 | ||
AK012567.1 | - | 4 | 497 | |||
AK012853.1 | - | 4 | 497 | |||
AK013523.1 | - | 4 | 507 | |||
AK014183.1 | - | 4 | 493 | |||
AK075962.1 | - | 4 | 495 | |||
AK160269.1 | - | 2 | 1104 | |||
AK208118.1 | - | 3 | 208 | |||
BC023306.1 | - | 4 | 471 | 4165774 |