Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk11ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027886.3 | NP_082162.3 | 12, 13 | 1278 | ||
GenBank mRNA | AK004757.2 | - | 12, 13 | 1231 | ||
AK156122.1 | - | 11, 12 | 1366 | |||
AK165911.1 | - | 12, 13 | 1269 | |||
AK171989.1 | - | 11, 12 | 1140 | |||
BC042403.1 | - | 10, 11 | 1315 | |||
BC061635.1 | - | 12, 13 | 1261 | 6530178 |