Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181328.3 | NP_851845.1 | 4 | 832 | ||
GenBank mRNA | AK047628.1 | - | 4 | 833 | ||
AK087057.1 | - | 4 | 820 | |||
AK148350.1 | - | 4 | 812 | |||
AK160906.1 | - | 4 | 782 | |||
BC006711.1 | - | 4 | 705 | 3584570 |