Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290321.2 | NP_001277250.1 | 44 | 10419 | ||
NM_001290322.2 | NP_001277251.1 | 43 | 11664 | |||
NM_005509.5 | NP_005500.4 | 43 | 10356 | |||
XM_005271909.4 | XP_005271966.1 | 46 | 10231 | |||
XM_005271910.4 | XP_005271967.1 | 44 | 10513 | |||
XM_005271912.2 | XP_005271969.1 | 46 | 10215 | |||
XM_011543212.2 | XP_011541514.1 | 45 | 10576 | |||
XM_011543213.2 | XP_011541515.1 | 46 | 10161 | |||
XM_011543214.2 | XP_011541516.1 | 42 | 9742 | |||
XM_011543215.2 | XP_011541517.1 | 41 | 9578 | |||
XM_017009141.1 | XP_016864630.1 | 45 | 10117 | |||
XM_017009142.1 | XP_016864631.1 | 44 | 9984 | |||
XM_017009143.1 | XP_016864632.1 | 47 | 10410 | |||
XM_017009144.1 | XP_016864633.1 | 45 | 10113 | |||
XM_017009145.1 | XP_016864634.1 | 45 | 9983 | |||
XM_017009146.1 | XP_016864635.1 | 45 | 11425 | |||
XM_017009148.1 | XP_016864637.1 | 44 | 10379 | |||
GenBank mRNA | AB593142.1 | - | 43 | 11667 | ||
AJ005821.1 | - | 43 | 9954 | |||
AK126726.1 | - | 5 | 2477 | |||
BX649064.1 | - | 11 | 2317 |