Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201226.1 | NP_957678.1 | 2 | 651 | ||
GenBank mRNA | AK129306.1 | - | 2 | 460 | ||
AK154897.1 | - | 2 | 681 | |||
BC094573.1 | - | 2 | 641 | 30354494 | ||
BC138462.1 | - | 2 | 723 | 8861482 | ||
BC138463.1 | - | 2 | 723 | 8861483 | ||
BU148242.1 | - | 2 | 645 |