Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027144.2 | NP_081420.2 | 10, 11 | 1299 | ||
GenBank mRNA | AF467766.1 | - | 10, 11 | 1064 | ||
AK122267.1 | - | 10, 11 | 1299 |