Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028544.1 | NP_082820.1 | 8, 9 | 2183 | ||
GenBank mRNA | AK011557.2 | - | 2, 3 | 266 | ||
AK033276.1 | - | 6, 7 | 1471 | |||
AK156602.1 | - | 8, 9 | 2195 | |||
BC028444.1 | - | 255 | ||||
BC072584.1 | - | 5, 6 | 1137 | |||
BC127059.1 | - | 6, 7 | 1109 | 40109852 | ||
BC150766.1 | - | 8, 9 | 2183 | 9087677 |