Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028544.1 | NP_082820.1 | 12 | 2979 | ||
GenBank mRNA | AK011557.2 | - | 6 | 1062 | ||
AK033276.1 | - | 10 | 2267 | |||
AK156602.1 | - | 12 | 2991 | |||
BC072584.1 | - | 9 | 1933 | |||
BC127059.1 | - | 10 | 1905 | 40109852 | ||
BC150766.1 | - | 12 | 2979 | 9087677 |