Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177919.2 | NP_808587.1 | 3 | 923 | ||
GenBank mRNA | AK032140.1 | - | 3 | 925 | ||
BC100415.1 | - | 3 | 948 | 6535676 |