Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spen | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019763.2 | NP_062737.2 | 3 | 782 | ||
GenBank mRNA | AB055980.1 | - | 2 | 227 | ||
AF156529.1 | - | 2 | 287 | |||
AK137647.1 | - | 3 | 670 |