Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324069.1 | NP_001310998.1 | 5 | 477 | ||
NM_001324072.1 | NP_001311001.1 | 4 | 256 | |||
NM_001324074.1 | NP_001311003.1 | 3 | 356 | |||
NM_001324075.1 | NP_001311004.1 | 4 | 390 | |||
NM_001324077.1 | NP_001311006.1 | 4 | 256 | |||
NM_001324078.1 | NP_001311007.1 | 3 | 356 | |||
NM_001799.3 | NP_001790.1 | 4 | 390 | |||
NR_136690.1 | 4 | 390 | ||||
GenBank mRNA | AK026509.1 | - | 4 | 275 | ||
AK308493.1 | - | 4 | 325 | |||
BC000834.2 | - | 4 | 229 | 3455907 | ||
BC005298.1 | - | 4 | 297 | 3997991 | ||
BI520184.1 | - | 4 | 390 | |||
BU660651.1 | - | 4 | 219 | |||
KU177970.1 | - | 4 | 209 | |||
KU177971.1 | - | 3 | 175 | |||
L20320.1 | - | 5 | 384 | |||
X77303.1 | - | 4 | 252 | |||
X77743.1 | - | 4 | 243 | |||
X79193.1 | - | 4 | 299 | |||
Y13120.1 | - | 4 | 312 |