Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164573.1 | NP_001158045.1 | 30 | 2985 | ||
GenBank mRNA | AK014505.1 | - | 8 | 1028 | ||
BC144867.1 | - | 9 | 890 | 9053415 | ||
BC144868.1 | - | 9 | 918 | |||
BC144869.1 | - | 9 | 818 | |||
BC144870.1 | - | 9 | 866 | 9053418 | ||
BC144872.1 | - | 7 | 947 |