Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpcat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173014.1 | NP_766602.1 | 14 | 2637 | ||
GenBank mRNA | AK039431.1 | - | 14 | 2637 |