Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppargc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008904.2 | NP_032930.1 | 13 | 4941 | ||
GenBank mRNA | AK032149.1 | - | 1 | 1411 | ||
AK043827.1 | - | 1 | 1445 |