Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144887.4 | NP_659136.1 | 6, 7 | 1901 | ||
GenBank mRNA | AK028737.1 | - | 3, 4 | 467 | ||
AK082513.1 | - | 6, 7 | 1826 | |||
AK145086.1 | - | 6, 7 | 1828 | |||
AK173251.1 | - | 6, 7 | 1856 | |||
AY894891.1 | - | 6, 7 | 1893 | |||
BC020051.1 | - | 5, 6 | 871 | 3987696 | ||
BC027047.1 | - | 2, 3 | 135 | |||
BC065155.1 | - | 6, 7 | 1771 | 5706087 |