Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144987.1 | NP_001138459.1 | 6 | 1702 | ||
NM_001144988.1 | NP_001138460.1 | 5 | 1315 | |||
NM_144895.2 | NP_659144.1 | 6 | 1486 | |||
GenBank mRNA | AK031545.1 | - | 5 | 1041 | ||
AK035081.1 | - | 4 | 1174 | |||
AK036569.1 | - | 4 | 638 | |||
AK087388.1 | - | 6 | 1455 | |||
AK089946.1 | - | 6 | 1487 | |||
AK129171.1 | - | 5 | 1294 | |||
AK147760.1 | - | 6 | 1704 | |||
AK159951.1 | - | 6 | 1487 | |||
AK192376.1 | - | 4 | 388 | |||
AK196543.1 | - | 3 | 223 | |||
AK200807.1 | - | 3 | 222 | |||
BC022921.1 | - | 6 | 1454 | 4220506 |