Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1l5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113416.1 | NP_001106887.1 | 9 | 879 | ||
NM_145506.4 | NP_663481.2 | 9 | 879 | |||
GenBank mRNA | AK034676.1 | - | 9 | 801 | ||
AK037024.1 | - | 9 | 835 | |||
AK040354.1 | - | 9 | 847 | |||
AK044959.1 | - | 9 | 817 | |||
AK044996.1 | - | 9 | 825 | |||
AK047523.1 | - | 9 | 806 | |||
AK079301.1 | - | 9 | 830 | |||
AK136125.1 | - | 9 | 834 | |||
AK161141.1 | - | 9 | 830 | |||
AK167449.1 | - | 9 | 879 | |||
AK173198.1 | - | 9 | 813 | |||
BC003937.1 | - | 9 | 864 | 3486543 | ||
BC011476.1 | - | 9 | 813 | 4014009 |