Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172684.2 | NP_766272.2 | 4 | 1890 | ||
GenBank mRNA | AB101658.1 | - | 4 | 1892 | ||
AK087685.1 | - | 3 | 856 | |||
AK122577.1 | - | 4 | 1425 | |||
AK134093.1 | - | 5 | 1009 | |||
AK138603.1 | - | 4 | 1434 | |||
BC094667.1 | - | 4 | 1415 | |||
BC139213.1 | - | 4 | 1859 | 8862235 | ||
BC139215.1 | - | 4 | 1859 | 8862237 |