Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177606.4 | NP_808274.2 | 5, 6 | 505 | ||
GenBank mRNA | AK029252.1 | - | 5, 6 | 507 | ||
AK173337.1 | - | 2, 3 | 161 | |||
BC021518.1 | - | 5, 6 | 477 | |||
BC125583.1 | - | 5, 6 | 470 | 40130098 | ||
BC137804.1 | - | 5, 6 | 467 | 8860823 |