Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177606.4 | NP_808274.2 | 7 | 644 | ||
GenBank mRNA | AK029252.1 | - | 7 | 646 | ||
AK173337.1 | - | 4 | 300 | |||
BC021518.1 | - | 7 | 616 | |||
BC125583.1 | - | 7 | 609 | 40130098 | ||
BC137804.1 | - | 7 | 606 | 8860823 |