Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021371.2 | NP_067346.1 | 6 | 1758 | ||
NM_181045.1 | NP_851388.1 | 7 | 1458 | |||
GenBank mRNA | AB041539.1 | - | 6 | 1749 | ||
AF282251.1 | - | 7 | 1440 | |||
AK081011.1 | - | 6 | 1209 | |||
AK158641.1 | - | 6 | 1120 | |||
AK158892.1 | - | 6 | 1730 | |||
AK163648.1 | - | 6 | 1221 | |||
BC057129.1 | - | 6 | 1737 | 6811387 | ||
BC058963.1 | - | 6 | 1737 | 6811387 |