Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138662.1 | NP_619603.1 | 1 | 1003 | ||
GenBank mRNA | AK129120.1 | - | 1 | 1105 | ||
AY013761.1 | - | 1 | 1003 | |||
BC141348.1 | - | 1 | 1040 | 9055960 |